2-Methyl-2H-indazole-5-carboxylic acid methyl ester 97+%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 2-methyl-2H-indazole-5-carboxylate

OR30723-250MG
APOSOR30723-250MGEA 33.5 EUR
APOSOR30723-250MG
2-Methyl-2H-indazole-5-carboxylic acid methyl ester 97+%
2-Methyl-2H-indazole-5-carboxylic acid methyl ester
Formel: C₁₀H₁₀N₂O₂
Molecular Weight: 190,2 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11109406
CAS-Nummer: 1092351-86-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR