2-(3,4-Dimethoxypyridin-2-yl)-3,4-dimethoxypyridine

Lieferant: Apollo Scientific
Achtung

OR307231-500MG
APOSOR307231-500MGEA 375 EUR
APOSOR307231-500MG
2-(3,4-Dimethoxypyridin-2-yl)-3,4-dimethoxypyridine
2-(3,4-Dimethoxypyridin-2-yl)-3,4-dimethoxypyridine
Formel: C₁₄H₁₆N₂O₄
Molecular Weight: 276,288 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD25976502
CAS-Nummer: 101664-55-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR