2-Chloro-3-methoxypyridin-4-ol

Lieferant: Apollo Scientific
Achtung

OR307236-100MG
APOSOR307236-100MGEA 21.9 EUR
APOSOR307236-100MG
2-Chloro-3-methoxypyridin-4-ol
2-Chloro-3-methoxypyridin-4-ol
Formel: C₆H₆ClNO₂
Molecular Weight: 159,57 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11553129
CAS-Nummer: 1227600-20-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR