2-Cyclopropylbenzeneboronic acid

Lieferant: Apollo Scientific
Achtung

OR307248-1G OR307248-5G OR307248-250MG
APOSOR307248-1GEA 0 EUR
APOSOR307248-1G APOSOR307248-5G APOSOR307248-250MG
2-Cyclopropylbenzeneboronic acid
2-Cyclopropylbenzeneboronic acid
Formel: C₉H₁₁BO₂
Lagertemperatur: Kühlschrank
MDL: MFCD12760762
CAS-Nummer: 1373393-41-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR