3-Cyclopropylbenzeneboronic acid

Lieferant: Apollo Scientific
Achtung

OR307249-100MG OR307249-1G OR307249-5G
APOSOR307249-100MGEA 161 EUR
APOSOR307249-100MG APOSOR307249-1G APOSOR307249-5G
3-Cyclopropylbenzeneboronic acid
3-Cyclopropylbenzeneboronic acid
Formel: C₉H₁₁BO₂
Lagertemperatur: Kühlschrank
MDL: MFCD11110340
CAS-Nummer: 1049730-10-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR