2-Cyclopropylbenzaldehyde 95+%

Lieferant: Apollo Scientific

OR307251-1G
APOSOR307251-1GEA 429 EUR
APOSOR307251-1G
2-Cyclopropylbenzaldehyde 95+%
2-Cyclopropylbenzaldehyde
Formel: C₁₀H₁₀O
Molecular Weight: 146.19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00480888
CAS-Nummer: 20034-51-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR