4-Cyclopropylbenzaldehyde

Lieferant: Apollo Scientific
Achtung

OR307253-1G
APOSOR307253-1GEA 250 EUR
APOSOR307253-1G
4-Cyclopropylbenzaldehyde
4-Cyclopropylbenzaldehyde
Formel: C₁₀H₁₀O
Flammpunkt: >70 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06802398
CAS-Nummer: 20034-50-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR