4-Bromo-3-iodo-1H-pyrazole
Lieferant: Apollo Scientific
|
Achtung
|
OR307260-1G
OR307260-250MG
OR307260-5G
APOSOR307260-1GEA
224
EUR
APOSOR307260-1G
APOSOR307260-250MG
APOSOR307260-5G
4-Bromo-3-iodo-1H-pyrazole
4-Bromo-3-iodo-1H-pyrazole
Formel:
C₃H₂BrIN₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD28975225 CAS-Nummer: 27258-14-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...