4-Bromo-3-iodo-1H-pyrazole

Lieferant: Apollo Scientific
Achtung

OR307260-1G OR307260-250MG OR307260-5G
APOSOR307260-1GEA 224 EUR
APOSOR307260-1G APOSOR307260-250MG APOSOR307260-5G
4-Bromo-3-iodo-1H-pyrazole
4-Bromo-3-iodo-1H-pyrazole
Formel: C₃H₂BrIN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD28975225
CAS-Nummer: 27258-14-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR