1-(Tetrahydro-2H-pyran-2-yl)-1H-indazole-4-carboxylic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(4-Carboxy-1H-indazol-1-yl)tetrahydro-2H-pyran, 4-Carboxy-1-(tetrahydro-2H-pyran-2-yl)-1H-indazole

OR30727-250MG
APOSOR30727-250MGEA 120 EUR
APOSOR30727-250MG
1-(Tetrahydro-2H-pyran-2-yl)-1H-indazole-4-carboxylic acid
1-(Tetrahydro-2H-pyran-2-yl)-1H-indazole-4-carboxylic acid
Formel: C₁₃H₁₄N₂O₃
Molecular Weight: 246,262 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09870059
CAS-Nummer: 1000576-23-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR