tert Butyl 3-iodo-1H-pyrazole-1-carboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: tert-Butyl 3-iodo-1H-pyrazole-1-carboxylate

OR307292-1G
APOSOR307292-1GEA 87 EUR
APOSOR307292-1G
tert Butyl 3-iodo-1H-pyrazole-1-carboxylate
tert Butyl 3-iodo-1H-pyrazole-1-carboxylate
Formel: C₈H₁₁IN₂O₂
Molecular Weight: 294.09 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD12407121
CAS-Nummer: 1233513-13-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR