Ethyl 1-(1-ethoxyethyl)-3-iodo-1H-pyrazole-4-carboxylate

Lieferant: Apollo Scientific
Achtung

OR307295-250MG
APOSOR307295-250MGEA 30.6 EUR
APOSOR307295-250MG
Ethyl 1-(1-ethoxyethyl)-3-iodo-1H-pyrazole-4-carboxylate
Ethyl 1-(1-ethoxyethyl)-3-iodo-1H-pyrazole-4-carboxylate
Formel: C₁₀H₁₅IN₂O₃
Molecular Weight: 338.142 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD32203993
CAS-Nummer: 1639858-75-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR