3-Iodo-4-methyl-1H-pyrazole

Lieferant: Apollo Scientific
Achtung

OR307296-1G
APOSOR307296-1GEA 270 EUR
APOSOR307296-1G
3-Iodo-4-methyl-1H-pyrazole
3-Iodo-4-methyl-1H-pyrazole
Formel: C₄H₅IN₂
Molecular Weight: 208 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20230556
CAS-Nummer: 24086-18-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR