1-(1-Ethoxyethyl)-3,4-diiodo-1H-pyrazole
Lieferant: Apollo Scientific
|
Achtung
|
OR307297-1G
APOSOR307297-1GEA
30.6
EUR
APOSOR307297-1G
1-(1-Ethoxyethyl)-3,4-diiodo-1H-pyrazole
1-(1-Ethoxyethyl)-3,4-diiodo-1H-pyrazole
Formel:
C₇H₁₀I₂N₂O Molecular Weight: 391,976 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD30721064 CAS-Nummer: 1639858-70-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...