4-Chloro-1-(1-ethoxyethyl)-3-iodo-1H-pyrazole

Lieferant: Apollo Scientific
Achtung

OR307300-1G OR307300-250MG
APOSOR307300-1GEA 364 EUR
APOSOR307300-1G APOSOR307300-250MG
4-Chloro-1-(1-ethoxyethyl)-3-iodo-1H-pyrazole
4-Chloro-1-(1-ethoxyethyl)-3-iodo-1H-pyrazole
Formel: C₇H₁₀ClIN₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD31537585
CAS-Nummer: 2197054-81-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR