1-(1-Ethoxyethyl)-3-iodo-1H-pyrazole

Lieferant: Apollo Scientific
Achtung

OR307301-1G OR307301-5G
APOSOR307301-1GEA 383 EUR
APOSOR307301-1G APOSOR307301-5G
1-(1-Ethoxyethyl)-3-iodo-1H-pyrazole
1-(1-Ethoxyethyl)-3-iodo-1H-pyrazole
Formel: C₇H₁₁IN₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD30721063
CAS-Nummer: 1639858-69-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR