1-(1-Ethoxyethyl)-4-iodo-1H-pyrazole-5-carbaldehyde

Lieferant: Apollo Scientific
Achtung

OR307304-1G
APOSOR307304-1GEA 192 EUR
APOSOR307304-1G
1-(1-Ethoxyethyl)-4-iodo-1H-pyrazole-5-carbaldehyde
1-(1-Ethoxyethyl)-4-iodo-1H-pyrazole-5-carbaldehyde
Formel: C₈H₁₁IN₂O₂
Molecular Weight: 294,09 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30721062
CAS-Nummer: 1639858-85-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR