tert-Butyl-5-amino-1H-indazole-1-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Amino-1H-indazole-1-carboxylic acid, 1,1 dimethyl ethyl ester, tert-Butyl 5-amino-1H-indazole-1-carboxylate, 1-Boc-5-amino-indazole, 5-Amino-indazole-1-carboxylic acid tert-butyl ester, N-(1)-Boc-5-aminoindazole
OR30732-100MG
APOSOR30732-100MGEA
21.9
EUR
APOSOR30732-100MG
tert-Butyl-5-amino-1H-indazole-1-carboxylate
tert-Butyl-5-amino-1H-indazole-1-carboxylate
Formel:
C₁₂H₁₅N₃O₂ Molecular Weight: 233,27 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD04114656 CAS-Nummer: 129488-10-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...