6-Brom-5-methyl-1H-indazol

Lieferant: Apollo Scientific
Achtung

OR30743-1G
APOSOR30743-1GEA 81.2 EUR
APOSOR30743-1G
6-Brom-5-methyl-1H-indazol
6-Bromo-5-methyl-1H-indazole
Formel: C₈H₇BrN₂
Lagertemperatur: Kühlschrank
MDL: MFCD09026997
CAS-Nummer: 1000343-69-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR