2-Chlor-6-methoxychinolin

Lieferant: Apollo Scientific
Achtung

OR307500-1G
APOSOR307500-1GEA 104 EUR
APOSOR307500-1G
2-Chlor-6-methoxychinolin
2-Chloro-6-methoxyquinoline
Formel: C₁₀H₈ClNO
Molecular Weight: 193,63 g/mol
Schmelzpunkt: 108…109 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00799212
CAS-Nummer: 13676-02-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR