4,7-Dibromoquinoline

Lieferant: Apollo Scientific

Synonyme: 4,7-Dibromo-1-azanaphthalene

OR307502-1G
APOSOR307502-1GEA 296 EUR
APOSOR307502-1G
4,7-Dibromoquinoline
4,7-Dibromoquinoline
Formel: C₉H₅Br₂N
Molecular Weight: 286,951 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08063188
CAS-Nummer: 700871-88-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR