8-Brom-4-chlorchinolin 97%

Lieferant: Apollo Scientific

OR307505-1G
APOSOR307505-1GEA 168 EUR
APOSOR307505-1G
8-Brom-4-chlorchinolin 97%
8-Bromo-4-chloroquinoline
Formel: C₉H₅BrClN
Molecular Weight: 242,4997 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01861852
CAS-Nummer: 65340-71-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR