4-Brom-7-chlorchinolin

Lieferant: Apollo Scientific

OR307507-5G
APOSOR307507-5GEA 121 EUR
APOSOR307507-5G
4-Brom-7-chlorchinolin
4-Bromo-7-chloroquinoline
Formel: C₉H₅BrClN
Lagertemperatur: Raumtemperatur
MDL: MFCD07700232
CAS-Nummer: 98519-65-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR