7-Bromo-2-chloro-4-methylquinoline
Lieferant: Apollo Scientific
OR307508-1G
APOSOR307508-1GEA
0
EUR
APOSOR307508-1G
7-Bromo-2-chloro-4-methylquinoline
7-Bromo-2-chloro-4-methylquinoline
Formel:
C₁₀H₇BrClN Lagertemperatur: Raumtemperatur |
MDL:
MFCD21647757 CAS-Nummer: 89446-52-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...