4-Brom-6-chlorchinolin
Lieferant: Apollo Scientific
OR307509-1G
APOSOR307509-1GEA
178
EUR
APOSOR307509-1G
4-Brom-6-chlorchinolin
4-Bromo-6-chloroquinoline
Formel:
C₉H₅BrClN Molecular Weight: 242,5000 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11505112 CAS-Nummer: 1070879-30-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...