4-Bromo-7,8-dichloroquinoline

Lieferant: Apollo Scientific

OR307511-1G
APOSOR307511-1GEA 498 EUR
APOSOR307511-1G
4-Bromo-7,8-dichloroquinoline
4-Bromo-7,8-dichloroquinoline
Formel: C₉H₄BrCl₂N
Lagertemperatur: Raumtemperatur
MDL: MFCD11505123
CAS-Nummer: 1070879-40-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR