7-Bromo-2-chloro-4,8-dimethylquinoline

Lieferant: Apollo Scientific

OR307512-1G
APOSOR307512-1GEA 0 EUR
APOSOR307512-1G
7-Bromo-2-chloro-4,8-dimethylquinoline
7-Bromo-2-chloro-4,8-dimethylquinoline
Formel: C₁₁H₉BrClN
Lagertemperatur: Raumtemperatur
MDL: MFCD21647771
CAS-Nummer: 1367706-81-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR