4-Bromo-3-chloroquinoline

Lieferant: Apollo Scientific

Synonyme: 4-Bromo-3-chloro-1-azanaphthalene

OR307513-1G
APOSOR307513-1GEA 556 EUR
APOSOR307513-1G
4-Bromo-3-chloroquinoline
4-Bromo-3-chloroquinoline
Formel: C₉H₅BrClN
Molecular Weight: 242,5 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13619679
CAS-Nummer: 1209339-16-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR