7-Bromo-3,4-dichloroquinoline

Lieferant: Apollo Scientific

OR307514-1G
APOSOR307514-1GEA 491 EUR
APOSOR307514-1G
7-Bromo-3,4-dichloroquinoline
7-Bromo-3,4-dichloroquinoline
Formel: C₉H₄BrCl₂N
Lagertemperatur: Raumtemperatur
MDL: MFCD13192969
CAS-Nummer: 1021435-01-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR