2-Amino-7-methoxy-3-methylquinoline hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: 7-Methoxy-3-methylquinolin-2-amine hydrochloride

OR307523-5G
APOSOR307523-5GEA 293 EUR
APOSOR307523-5G
2-Amino-7-methoxy-3-methylquinoline hydrochloride
2-Amino-7-methoxy-3-methylquinoline hydrochloride
Formel: C₁₁H₁₂N₂O·ClH
Molecular Weight: 224,687 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10703342
CAS-Nummer: 1171863-09-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR