6-Bromo-2-hydrazino-3-methylquinoline hydrochloride
Lieferant: Apollo Scientific
Synonyme:
6-Bromo-2-hydrazino-3-methyl-1-azanaphthalene hydrochloride
OR307524-1G
APOSOR307524-1GEA
383
EUR
APOSOR307524-1G
6-Bromo-2-hydrazino-3-methylquinoline hydrochloride
6-Bromo-2-hydrazino-3-methylquinoline hydrochloride
Formel:
C₁₀H₁₀BrN₃·ClH Molecular Weight: 288,571 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11505170 CAS-Nummer: 1172069-28-1 |
Learn more
![](/m-de.vwr.com/de_DE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...