7-Bromo-2-hydrazino-3-methylquinoline hydrochloride

Lieferant: Apollo Scientific

Synonyme: 7-Bromo-2-hydrazino-3-methyl-1-azanaphthalene hydrochloride

OR307526-1G
APOSOR307526-1GEA 491 EUR
APOSOR307526-1G
7-Bromo-2-hydrazino-3-methylquinoline hydrochloride
7-Bromo-2-hydrazino-3-methylquinoline hydrochloride
Formel: C₁₀H₁₀BrN₃·ClH
Molecular Weight: 288.571 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11505171
CAS-Nummer: 1171639-07-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR