cis-6-Amino-cyclohex-3-enecarboxylic acid
Lieferant: Apollo Scientific
OR307528-1G
APOSOR307528-1GEA
451
EUR
APOSOR307528-1G
cis-6-Amino-cyclohex-3-enecarboxylic acid
cis-6-Amino-cyclohex-3-enecarboxylic acid
Formel:
C₇H₁₁NO₂ Molecular Weight: 141,168 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00143983 CAS-Nummer: 54162-90-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...