3-Amino-4-ethylcapronsäure
Lieferant: Apollo Scientific
OR307535-1G
APOSOR307535-1GEA
285
EUR
APOSOR307535-1G
3-Amino-4-ethylcapronsäure
3-Amino-4-ethylcapronsäure
Formel:
C₈H₁₇NO₂ Molecular Weight: 159,23 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01863280 CAS-Nummer: 204191-42-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...