3-Amino-3-(3,4-dichlorphenyl)propionsäure

Lieferant: Apollo Scientific

Synonyme: 3-Amino-3-(3,4-dichlorophenyl)propanoic acid

OR307537-1G
APOSOR307537-1GEA 503 EUR
APOSOR307537-1G
3-Amino-3-(3,4-dichlorphenyl)propionsäure
3-Amino-3-(3,4-dichlorphenyl)propionsäure
Formel: C₉H₉Cl₂NO₂
Molecular Weight: 234,0793 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00462884
CAS-Nummer: 117391-57-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR