3-Amino-5-phenyl-pentanoic acid

Lieferant: Apollo Scientific

OR307539-250MG
APOSOR307539-250MGEA 127 EUR
APOSOR307539-250MG
3-Amino-5-phenyl-pentanoic acid
3-Amino-5-phenyl-pentanoic acid
Formel: C₁₁H₁₅NO₂
Molecular Weight: 193.242 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01318914
CAS-Nummer: 91247-38-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR