(6,7-Dimethoxy-1,2,3,4-tetrahydro-isoquinolin-1-yl)-acetic acid

Lieferant: Apollo Scientific

OR307542-1G
APOSOR307542-1GEA 317 EUR
APOSOR307542-1G
(6,7-Dimethoxy-1,2,3,4-tetrahydro-isoquinolin-1-yl)-acetic acid
(6,7-Dimethoxy-1,2,3,4-tetrahydro-isoquinolin-1-yl)-acetic acid
Formel: C₁₃H₁₇NO₄
Molecular Weight: 251,278 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00835612
CAS-Nummer: 68345-67-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR