(6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinolin-1-yl)acetonitrile
Lieferant: Apollo Scientific
OR307544-1G
APOSOR307544-1GEA
383
EUR
APOSOR307544-1G
(6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinolin-1-yl)acetonitrile
(6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinolin-1-yl)acetonitrile
Formel:
C₁₃H₁₆N₂O₂ Molecular Weight: 232.28 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00800509 CAS-Nummer: 52244-06-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...