Boc-(1R,2S)-(-)-2-aminocyclohex-4-enecarboxylic acid

Lieferant: Apollo Scientific

OR307545-1G
APOSOR307545-1GEA 151 EUR
APOSOR307545-1G
Boc-(1R,2S)-(-)-2-aminocyclohex-4-enecarboxylic acid
Boc-(1R,2S)-(-)-2-aminocyclohex-4-enecarboxylic acid
Formel: C₁₂H₁₉NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD09750522
CAS-Nummer: 63216-52-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR