trans-6-tert-Butoxycarbonylamino-cyclohex-3-enecarboxylic acid

Lieferant: Apollo Scientific
Achtung

OR307546-1G
APOSOR307546-1GEA 201 EUR
APOSOR307546-1G
trans-6-tert-Butoxycarbonylamino-cyclohex-3-enecarboxylic acid
trans-6-tert-Butoxycarbonylamino-cyclohex-3-enecarboxylic acid
Formel: C₁₂H₁₉NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD02682629
CAS-Nummer: 865689-24-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR