cis-6-Amino-cyclohex-3-enecarboxylic acid amide hydrochloride

Lieferant: Apollo Scientific

OR307549-1G
APOSOR307549-1GEA 498 EUR
APOSOR307549-1G
cis-6-Amino-cyclohex-3-enecarboxylic acid amide hydrochloride
cis-6-Amino-cyclohex-3-enecarboxylic acid amide hydrochloride
Formel: C₇H₁₂N₂O·ClH
Molecular Weight: 176,644 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20036289
CAS-Nummer: 1980007-32-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR