diendo-3-Benzoyl-bicyclo[2.2.1]heptane-2-carboxylic acid

Lieferant: Apollo Scientific

OR307554-1G
APOSOR307554-1GEA 219 EUR
APOSOR307554-1G
diendo-3-Benzoyl-bicyclo[2.2.1]heptane-2-carboxylic acid
diendo-3-Benzoyl-bicyclo[2.2.1]heptane-2-carboxylic acid
Formel: C₁₅H₁₆O₃
Molecular Weight: 244,286 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09260353
CAS-Nummer: 212757-12-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR