diexo-3-Amino-7-oxa-bicyclo[2.2.1]hept-5-ene-2-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR307560-250MG
APOSOR307560-250MGEA 213 EUR
APOSOR307560-250MG
diexo-3-Amino-7-oxa-bicyclo[2.2.1]hept-5-ene-2-carboxylic acid
diexo-3-Amino-7-oxa-bicyclo[2.2.1]hept-5-ene-2-carboxylic acid
Formel: C₇H₉NO₃
Molecular Weight: 155,151 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07783748
CAS-Nummer: 783340-43-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR