diexo-3-Amino-7-oxa-bicyclo[2.2.1]heptane-2-carboxylic acid

Lieferant: Apollo Scientific
Gefahr

OR307561-1G
APOSOR307561-1GEA 517 EUR
APOSOR307561-1G
diexo-3-Amino-7-oxa-bicyclo[2.2.1]heptane-2-carboxylic acid
diexo-3-Amino-7-oxa-bicyclo[2.2.1]heptane-2-carboxylic acid
Formel: C₇H₁₁NO₃
Molecular Weight: 157.167 g/mol
Schmelzpunkt: 251…252 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07783750
CAS-Nummer: 4576-30-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR