1-Carboxymethyl-6,7-dimethoxy-3,4-dihydro-1H-isoquinoline-2-carboxylic acid tert-butyl ester
Lieferant: Apollo Scientific
OR307564-1G
OR307564-250MG
APOSOR307564-1GEA
525
EUR
APOSOR307564-1G
APOSOR307564-250MG
1-Carboxymethyl-6,7-dimethoxy-3,4-dihydro-1H-isoquinoline-2-carboxylic acid tert-butyl ester
1-Carboxymethyl-6,7-dimethoxy-3,4-dihydro-1H-isoquinoline-2-carboxylic acid tert-butyl ester
Formel:
C₁₈H₂₅NO₆ Molecular Weight: 351.394 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02258126 CAS-Nummer: 282524-92-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...