3-tert-Butoxycarbonylamino-pentanoic acid

Lieferant: Apollo Scientific

Synonyme: 3-((tert-Butoxycarbonyl)amino)pentanoic acid

OR307566-250MG
APOSOR307566-250MGEA 88.6 EUR
APOSOR307566-250MG
3-tert-Butoxycarbonylamino-pentanoic acid
3-tert-Butoxycarbonylamino-pentanoic acid
Formel: C₁₀H₁₉NO₄
Molecular Weight: 217,262 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05863633
CAS-Nummer: 557091-78-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR