3-Amino-hexanoic acid

Lieferant: Apollo Scientific

Synonyme: 3-Aminohexanoic acid

OR307567-1G
APOSOR307567-1GEA 839 EUR
APOSOR307567-1G
3-Amino-hexanoic acid
3-Amino-hexanoic acid
Formel: C₆H₁₃NO₂
Molecular Weight: 131,173 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00044473
CAS-Nummer: 58521-63-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR