3-tert-Butoxycarbonylamino-heptanoic acid

Lieferant: Apollo Scientific

OR307569-1G
APOSOR307569-1GEA 498 EUR
APOSOR307569-1G
3-tert-Butoxycarbonylamino-heptanoic acid
3-tert-Butoxycarbonylamino-heptanoic acid
Formel: C₁₂H₂₃NO₄
Molecular Weight: 245.315 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03412496
CAS-Nummer: 676348-90-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR