3-tert-Butoxycarbonylamino-5-phenyl-pentanoic acid

Lieferant: Apollo Scientific

OR307572-1G
APOSOR307572-1GEA 383 EUR
APOSOR307572-1G
3-tert-Butoxycarbonylamino-5-phenyl-pentanoic acid
3-tert-Butoxycarbonylamino-5-phenyl-pentanoic acid
Formel: C₁₆H₂₃NO₄
Molecular Weight: 293,358 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05863635
CAS-Nummer: 955314-80-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR