3-Amino-3-(4-isopropylphenyl)propionsäure

Lieferant: Apollo Scientific

OR307573-1G
APOSOR307573-1GEA 68.3 EUR
APOSOR307573-1G
3-Amino-3-(4-isopropylphenyl)propionsäure
3-Amino-3-(4-isopropylphenyl)propionsäure
Formel: C₁₂H₁₇NO₂
Molecular Weight: 207,2700 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02663184
CAS-Nummer: 117391-53-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR